Ammonium peroxodisulphate (APS), Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Peroxidisulphuric acid diammonium salt, Ammonium persulphate
M-Clarity™ quality level = MQ200
A7460-100G A7460-500G
SIALA7460-100GEA 208 EUR
SIALA7460-100G SIALA7460-500G
Ammonium peroxodisulphate (APS), Sigma-Aldrich®
Ammonium peroxodisulphate (APS)
Formula: (NH₄)₂S₂O₈
Melting Pt: 120 °C
Density: 1.98 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003390
CAS Number: 7727-54-0
EINECS: 231-786-5
UN: 1444
ADR: 5.1,III
Merck Index: 13,00545
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f82a373f21d34e63dc94c0598f3f33ba6bbfdb2b&vid=2b6d196dbf07ce4dd528ac7b4f8456678accd3ba&prid=web&clid=SAPDAM