Ammonium sulphate ≥98%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Sulphuric acid diammonium salt, Diammonium sulphate, Mascagnite
A11682.0B A11682.0I
A11682.0BEA 39 EUR
A11682.0B A11682.0I
Ammonium sulphate ≥98%
Ammonium sulphate
Purity: 98+% CAS No.: 7783-20-2 Molecular Formula: H8N2O4S Formula Weight: 132.14 Melting Point: ca 280° dec. Merck Reference: 14,555 MDL No.: MFCD00003391
Formula: (NH₄)₂SO₄
MW: 132.14 g/mol
Melting Pt: ∼ 280 °C (dec.)
Density: 1.769
Storage Temperature: Ambient
MDL Number: MFCD00003391
CAS Number: 7783-20-2
EINECS: 231-984-1
REACH: 01-2119455044-46
Merck Index: 13,00559