Ammonium sulphate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
A3920-1KG A3920-500G
SIALA3920-1KGEA 154 EUR
SIALA3920-1KG SIALA3920-500G
Ammonium sulphate, Sigma-Aldrich®
Ammonium sulphate
Formula: (NH₄)₂SO₄
Melting Pt: 336…339 °C
Density: 1.78 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003391
CAS Number: 7783-20-2
EINECS: 231-984-1
REACH: 01-2119455044-46
Merck Index: 13,00559