Chlorosulphonyl isocyanate 98+%
Supplier: Thermo Fisher Scientific
|
Danger
|
162040050
162040500
162042500
162040010
ACRO162040050EA
27.5
EUR
ACRO162040050
ACRO162040500
ACRO162042500
ACRO162040010
Chlorosulphonyl isocyanate 98+%
Chlorosulphonyl isocyanate
Formula:
CClNO₃S Boiling Pt: 106 °C (760 mmHg) Melting Pt: –44 °C Density: 1.626 g/cm³ Flash Pt: >110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00011608 CAS Number: 1189-71-5 EINECS: 214-715-2 UN: 3265 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4450 to 1.4490 (20°C, 589 nm) |
Titration with NaOH | ≥98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...