Sulphuryl chloride, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: Sulphuryl dichloride

M-Clarity™ quality level = MQ200

157767-1L 157767-25ML
SIAL157767-1LEA 149 EUR
SIAL157767-1L SIAL157767-25ML
Sulphuryl chloride, Sigma-Aldrich®
Sulphuryl chloride
Formula: Cl₂O₂S
Boiling Pt: 69.1 °C (1013 hPa)
Melting Pt: –54 °C
Density: 1.67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011451
CAS Number: 7791-25-5
EINECS: 232-245-6
UN: 1834
ADR: 8,I
Merck Index: 12,09153

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR