Diallyl sulphide, Sigma-Aldrich®
Supplier: Merck
Synonyms: Allyl sulphide
M-Clarity™ quality level = MQ300
W204218-100G-K W204218-1KG-K W204218-4KG-K
SIALW204218-100G-KEA 73.6 EUR
SIALW204218-100G-K SIALW204218-1KG-K SIALW204218-4KG-K
Diallyl sulphide, Sigma-Aldrich®
Diallyl sulphide
Formula: C₆H₁₀S
Boiling Pt: 139…140 °C (1013 hPa)
Melting Pt: –83 °C
Density: 0.89 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008658
CAS Number: 592-88-1
EINECS: 209-775-1
UN: 1993
ADR: 3,III
Merck Index: 13,00294