Dimethyl sulphoxide ≥99%
Supplier: Thermo Fisher Scientific
Warning
|
Synonyms:
Methanesulphinylmethane, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO
A13280.36
A13280.0C
A13280.0E
A13280.36EA
35.3
EUR
A13280.36
A13280.0C
A13280.0E
Dimethyl sulphoxide ≥99%
Dimethyl sulphoxide
Purity: 99+%
CAS No.: 67-68-5
Molecular Formula: C2H6OS
Formula Weight: 78.13
Flash Point: 87°(189°F)
Refractive Index: 1.4790
Odour: Mild
Melting Point: 18.4°
Boiling Point: 189°
Beilstein Reference: 506008
Merck Reference: 14,3259
MDL No.: MFCD00002089
Formula:
(CH₃)₂SO Boiling Pt: 189 °C Melting Pt: 18.4 °C Density: 1.101 Flash Pt: 87 °C (189 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002089 CAS Number: 67-68-5 EINECS: 200-664-3 Merck Index: 14,3259 |