Thiodiglycol, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
88561-1L 88561-2.5L
SIAL88561-1LEA 92.9 EUR
SIAL88561-1L SIAL88561-2.5L
Thiodiglycol, Sigma-Aldrich®
Thiodiglycol
Formula: S(CH₂CH₂OH)₂
Boiling Pt: 282 °C (1013 hPa)
Melting Pt: –10 °C
Density: 1.18 g/cm³ (20 °C)
Flash Pt: 113 °C (closed cup)
CAS Number: 111-48-8
EINECS: 203-874-3
Merck Index: 12,09466
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=28ad1d0271b8a40084a5753c4df7774ab45bb720&vid=6ff591bf6981e5a6f12e9bc3d4f57311542d56bb&prid=web&clid=SAPDAM