Dimethyl disulphide, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
Dimethyl disulphide, Methyl disulphide
M-Clarity™ quality level = MQ100
528013-1L
528013-25ML
SIAL528013-1LEA
73.1
EUR
SIAL528013-1L
SIAL528013-25ML
Dimethyl disulphide, Sigma-Aldrich®
Dimethyl disulphide
Formula:
C₂H₆S₂ Boiling Pt: 110 °C (1013 hPa) Melting Pt: –98 °C Density: 1.06 g/cm³ (20 °C) Flash Pt: 16 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008561 CAS Number: 624-92-0 EINECS: 210-871-0 UN: 2381 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...