Dibenzyl disulphide, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
Benzyl disulphide
M-Clarity™ quality level = MQ100
B21805-100G
B21805-500G
SIALB21805-100GEA
69
EUR
SIALB21805-100G
SIALB21805-500G
Dibenzyl disulphide, Sigma-Aldrich®
Dibenzyl disulphide
Formula:
C₁₄H₁₄S₂ Boiling Pt: 142…148 °C (0.05 mmHg) Melting Pt: 62…67 °C Density: 1.302 g/cm³ (25 °C) |
MDL Number:
MFCD00004783 CAS Number: 150-60-7 EINECS: 205-764-0 Merck Index: 13,03037 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...