Iodine monobromide, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ200
224847-250G 224847-50G
SIAL224847-250GEA 267 EUR
SIAL224847-250G SIAL224847-50G
Iodine monobromide, Sigma-Aldrich®
Iodine monobromide
Formula: IBr
MW: 206.81 g/mol
Boiling Pt: 116
Melting Pt: 37…40 °C
Density: 4.41 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011353
CAS Number: 7789-33-5
EINECS: 232-159-9
UN: 3260
ADR: 8 II
Merck Index: 13,05038