Iodine monochloride, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Iodochloride
M-Clarity™ quality level = MQ100
481556-25G 481556-5G
SIAL481556-25GEA 151 EUR
SIAL481556-25G SIAL481556-5G
Iodine monochloride, Sigma-Aldrich®
Iodine monochloride
Formula: ClI
MW: 162.36 g/mol
Boiling Pt: 97.3
Melting Pt: 26
Density: 3.1 g/cm³ (29 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011354
CAS Number: 7790-99-0
EINECS: 232-236-7
UN: 1792
ADR: 8 II
Merck Index: 13,05039