Imidazole, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: Glyoxaline, 1H-Imidazole, Glyoxalamine

M-Clarity™ quality level = MQ300

I0250-100G I0250-250G I0250-25G I0250-5G
SIALI0250-100GEA 455 EUR
SIALI0250-100G SIALI0250-250G SIALI0250-25G SIALI0250-5G
Imidazole, Sigma-Aldrich®
Imidazole
Formula: C₃H₄N₂
MW: 68.08 g/mol
Boiling Pt: 255
Melting Pt: 89…90 °C
Density: 1.18 g/cm³ (20 °C)
Flash Pt: 135
Storage Temperature: Ambient
MDL Number: MFCD00005183
CAS Number: 288-32-4
EINECS: 206-019-2
UN: 2923
ADR: 8 III
Merck Index: 13,04935
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48eba49e61201d6fffa2bef0c54633a20660c7ef&vid=e7f154e4976b52732539e49bbff60454d5eb82ff&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR