Imidazole, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ200
I202-100G I202-2KG I202-500G I202-5G
SIALI202-100GEA 92.2 EUR
SIALI202-100G SIALI202-2KG SIALI202-500G SIALI202-5G
Imidazole, Sigma-Aldrich®
Imidazole
Formula: C₃H₄N₂
Boiling Pt: 255 °C (1013 hPa)
Melting Pt: 89…90 °C
Density: 1.18 g/cm³ (20 °C)
Flash Pt: 135 °C
Storage Temperature: Ambient
MDL Number: MFCD00005183
CAS Number: 288-32-4
EINECS: 206-019-2
UN: 2923
ADR: 8,III
Merck Index: 13,04935
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48eba49e61201d6fffa2bef0c54633a20660c7ef&vid=e7f154e4976b52732539e49bbff60454d5eb82ff&prid=web&clid=SAPDAM