4-Methylimidazole ≥98%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: 4-Methyl-1H-imidazole
B24139.09 B24139.18 B24139.30
B24139.09EA 33.2 EUR
B24139.09 B24139.18 B24139.30
4-Methylimidazole ≥98%
4-Methylimidazole
Purity: 98% CAS No.: 822-36-6 Molecular Formula: C4H6N2 Formula Weight: 82.11 Flash Point: 157°(314°F) Melting Point: 49-57° Boiling Point: 263° Beilstein Reference: 1453 UN No.: 3263 MDL No.: MFCD00005201
Formula: C₄H₆N₂
MW: 82.11 g/mol
Boiling Pt: 263 °C
Melting Pt: 49…57 °C
Density: 1.020
Flash Pt: 157 °C (314 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005201
CAS Number: 822-36-6
EINECS: 212-497-3
UN: 3263
ADR: 8 III