Indene 90% stabilised, Technical Grade
Supplier: Thermo Fisher Scientific
Warning
122140010 122140050 122142500
ACRO122140010EA 209 EUR
ACRO122140010 ACRO122140050 ACRO122142500
Indene 90% stabilised, Technical Grade
Indene
Formula: C₉H₈
Boiling Pt: 181 °C (760 mmHg)
Melting Pt: 5…3 °C
Density: 0.99 g/cm³
Flash Pt: 56 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003777
CAS Number: 95-13-6
EINECS: 202-393-6
UN: 3295
ADR: 3,III
Merck Index: 13,04964
Order Now
Specification Test Results
Appearance Clear yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5670 to 1.5750 (20°C, 589 nm)
GC ≥88 %
Stabilizer 5 to 10 ppm tert-Butylcatechol