2-Methylindole, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 2-Methyl-1H-indole
M-Clarity™ quality level = MQ200
M51407-100G M51407-25G
SIALM51407-100GEA 80.7 EUR
SIALM51407-100G SIALM51407-25G
2-Methylindole, Sigma-Aldrich®
2-Methylindole
Formula: C₉H₉N
MW: 131.18 g/mol
Boiling Pt: 273
Melting Pt: 56…59 °C
Density: 1.07 g/cm³ (20 °C)
Flash Pt: 141
Storage Temperature: Refrigerator
MDL Number: MFCD00005616
CAS Number: 95-20-5
EINECS: 202-398-3