3-Methylindole, Sigma-Aldrich®
Supplier: Merck
Synonyms: Skatol, 3-Methyl-1H-indole, β-Methylindole, Skatole
M-Clarity™ quality level = MQ100
M51458-100G M51458-25G M51458-5G
SIALM51458-100GEA 312 EUR
SIALM51458-100G SIALM51458-25G SIALM51458-5G
3-Methylindole, Sigma-Aldrich®
3-Methylindole
Formula: C₉H₉N
MW: 131.18 g/mol
Boiling Pt: 265
Melting Pt: 95…97 °C
Density: 1.003 g/cm³ (99.6 °C)
MDL Number: MFCD00005627
CAS Number: 83-34-1
EINECS: 201-471-7
Merck Index: 12,08700