7-Methylindole, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
M51490-1G M51490-5G
SIALM51490-1GEA 27.2 EUR
SIALM51490-1G SIALM51490-5G
7-Methylindole, Sigma-Aldrich®
7-Methylindole
Formula: C₉H₉N
MW: 131.18 g/mol
Boiling Pt: 266
Melting Pt: 83
Density: 1.0202 g/cm³ (99.6 °C)
MDL Number: MFCD00005684
CAS Number: 933-67-5
EINECS: 213-270-1