Cerium trichloride, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: Cerous chloride, Cerium(III) chloride, Cerium (III) chloride

M-Clarity™ quality level = MQ100

429406-25G 429406-5G
SIAL429406-25GEA 443 EUR
SIAL429406-25G SIAL429406-5G
Cerium trichloride, Sigma-Aldrich®
Cerium trichloride
Formula: CeCl₃
MW: 246.47 g/mol
Melting Pt: 848
Density: 3.193 g/cm³ (20 °C)
MDL Number: MFCD00010929
CAS Number: 7790-86-5
EINECS: 232-227-8
Merck Index: 13,02008

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR