Europium(II) chloride, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

431850-1G 431850-5G
SIAL431850-1GEA 209 EUR
SIAL431850-1G SIAL431850-5G
Europium(II) chloride, Sigma-Aldrich®
Europium (II) chloride
Formula: Cl₂Eu
CAS Number: 13769-20-5
EINECS: 237-386-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR