Terbium(III) chloride, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

451304-10G 451304-1G
SIAL451304-10GEA 873 EUR
SIAL451304-10G SIAL451304-1G
Terbium(III) chloride, Sigma-Aldrich®
Terbium (III) chloride
Formula: Cl₃Tb
MW: 265.28 g/mol
Melting Pt: 588
Density: 4.35 g/cm³ (25 °C)
MDL Number: MFCD00011258
CAS Number: 10042-88-3
EINECS: 233-132-4
Merck Index: 13,09087

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR