Thulium(III) chloride, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
439649-1G
439649-5G
SIAL439649-1GEA
96.2
EUR
SIAL439649-1G
SIAL439649-5G
Thulium(III) chloride, Sigma-Aldrich®
Thulium (III) chloride
Formula:
Cl₃Tm MW: 275.29 g/mol Melting Pt: 820.85 |
MDL Number:
MFCD00011283 CAS Number: 13537-18-3 EINECS: 236-904-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...