Cerium(IV) oxide, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

202975-50G 202975-10G
SIAL202975-50GEA 583 EUR
SIAL202975-50G SIAL202975-10G
Cerium(IV) oxide, Sigma-Aldrich®
Cerium (IV) oxide
Formula: CeO₂
Boiling Pt: 3468 °C (1013 hPa)
Melting Pt: 2000 °C
Density: 7.13 g/cm³ (25 °C)
MDL Number: MFCD00010933
CAS Number: 1306-38-3
EINECS: 215-150-4
Merck Index: 12,02035

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR