Lanthanum(III) oxide, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
199923-100G
199923-500G
SIAL199923-100GEA
207
EUR
SIAL199923-100G
SIAL199923-500G
Lanthanum(III) oxide, Sigma-Aldrich®
Lanthanum (III) oxide
Formula:
La₂O₃ MW: 325.81 g/mol Boiling Pt: 4200 Melting Pt: 2315 Density: 6.51 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011071 CAS Number: 1312-81-8 EINECS: 215-200-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...