Neodymium(III) oxide, Sigma-Aldrich®
Supplier: Merck
Synonyms: Dineodymium trioxide, Neodymium(III)-oxide, Neodymium oxide
M-Clarity™ quality level = MQ100
634611-50G 634611-5G
SIAL634611-50GEA 619 EUR
SIAL634611-50G SIAL634611-5G
Neodymium(III) oxide, Sigma-Aldrich®
Neodymium (III) oxide
Formula: Nd₂O₃
MW: 336.48 g/mol
Boiling Pt: 3760
Melting Pt: 2270
Density: 7.24 g/cm³ (20 °C)
MDL Number: MFCD00011134
CAS Number: 1313-97-9
EINECS: 215-214-1
Merck Index: 14,06450