Terbium(III) oxide, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

590509-10G 590509-2G
SIAL590509-10GEA 578 EUR
SIAL590509-10G SIAL590509-2G
Terbium(III) oxide, Sigma-Aldrich®
Terbium (III) oxide
Formula: O₃Tb₂
MW: 365.85 g/mol
CAS Number: 12036-41-8
EINECS: 234-849-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR