Ammonium cerium(IV) nitrate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Ammonium ceric nitrate, Ammonium hexanitratocerate (IV), Ceric Ammonium Nitrate, Cerium (IV) ammonium nitrate, Diammonium hexanitratocerate
MQ200
229547-10G 229547-250G 229547-50G
SIAL229547-10GEA 55.7 EUR
SIAL229547-10G SIAL229547-250G SIAL229547-50G
Ammonium cerium(IV) nitrate, Sigma-Aldrich®
Ammonium cerium (IV) nitrate
Formula: (NH₄)₂Ce(NO₃)₆
MW: 548.22 g/mol
Melting Pt: 107…108 °C
Density: 2.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00151121
CAS Number: 16774-21-3
EINECS: 240-827-6
UN: 1477
ADR: 5.1 III
Merck Index: 13,00509