1-Naphthaldehyde, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: α-Naphthaldehyde
M-Clarity™ quality level = MQ200
N109-100G-A N109-2G-A N109-500G-A
SIALN109-100G-AEA 65.2 EUR
SIALN109-100G-A SIALN109-2G-A SIALN109-500G-A
1-Naphthaldehyde, Sigma-Aldrich®
1-Naphthaldehyde
Formula: C₁₁H₈O
MW: 156.18 g/mol
Boiling Pt: 292
Melting Pt: 2
Density: 1.14 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004003
CAS Number: 66-77-3
EINECS: 200-633-4