1,5-Diaminonaphthalene 97%
Supplier: Thermo Fisher Scientific
Warning
B24262.06 B24262.14 B24262.22
B24262.06EA 27.3 EUR
B24262.06 B24262.14 B24262.22
1,5-Diaminonaphthalene 97%
1,5-Diaminonaphthalene
Purity: 97% CAS No.: 2243-62-1 Molecular Formula: C10H10N2 Formula Weight: 158.20 Melting Point: 189-192° Boiling Point: 200-210°/5mm UN No.: 3077 MDL No.: MFCD00004029
Formula: C₁₀H₁₀N₂
Boiling Pt: 200…210 °C (5 mmHg)
Melting Pt: 189…192 °C
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004029
CAS Number: 2243-62-1
EINECS: 218-817-8
UN: 3077
ADR: 9,III