1,8-Diaminonaphthalene, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ200
D21405-100G D21405-5G
SIALD21405-100GEA 109 EUR
SIALD21405-100G SIALD21405-5G
1,8-Diaminonaphthalene, Sigma-Aldrich®
1,8-Diaminonaphthalene
Formula: C₁₀H₁₀N₂
Boiling Pt: 352 °C (1013 hPa)
Melting Pt: 64…65 °C
Density: 1.18 g/cm³ (20 °C)
Flash Pt: 195 °C
Storage Temperature: Ambient
MDL Number: MFCD00004033
CAS Number: 479-27-6
EINECS: 207-529-8
UN: 3259
ADR: 8,III
Merck Index: 12,06459