2-Ethoxynaphthalene ≥99%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Ethyl 2-naphthyl ether
B23327.14 B23327.22 B23327.36
B23327.14EA 22.7 EUR
B23327.14 B23327.22 B23327.36
2-Ethoxynaphthalene ≥99%
2-Ethoxynaphthalene
Purity: 99% CAS No.: 93-18-5 Molecular Formula: C12H12O Formula Weight: 172.23 Flash Point: 134°(273°F) Melting Point: 35-37° Boiling Point: 282° Beilstein Reference: 2042397 Merck Reference: 14,3752 MDL No.: MFCD00016808
Formula: C₁₂H₁₂O
MW: 172.23 g/mol
Boiling Pt: 282 °C
Melting Pt: 35…37 °C
Density: 1.064 g/cm³ (20 °C)
Flash Pt: 134 °C (273 °F)
Storage Temperature: Ambient
MDL Number: MFCD00016808
CAS Number: 93-18-5
EINECS: 202-226-7
Merck Index: 13,03788