2-Naphthol, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
2-Hydroxynaphthalene, β-Naphthol, β-Hydroxynaphthalene
M-Clarity™ quality level = MQ200
185507-100G
185507-500G
185507-5G
SIAL185507-100GEA
37.7
EUR
SIAL185507-100G
SIAL185507-500G
SIAL185507-5G
2-Naphthol, Sigma-Aldrich®
2-Naphthol
Formula:
C₁₀H₈O Boiling Pt: 286 °C (1013 hPa) Melting Pt: 121.6 °C Density: 1.217 g/cm³ (20 °C) Flash Pt: 153 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004067 CAS Number: 135-19-3 EINECS: 205-182-7 UN: 3077 ADR: 9,III Merck Index: 12,06471 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...