1,5-Dihydroxynaphthalene 97%
Supplier: Thermo Fisher Scientific
Warning
115001000 115005000
ACRO115001000EA 40.5 EUR
ACRO115001000 ACRO115005000
1,5-Dihydroxynaphthalene 97%
1,5-Dihydroxynaphthalene
Formula: C₁₀H₈O₂
Boiling Pt: 375 °C (1013 hPa)
Melting Pt: 259…261 °C (dec.)
Density: 1,41 g/cm³ (20 °C)
Flash Pt: 252 °C
Storage Temperature: Ambient
MDL Number: MFCD00003980
CAS Number: 83-56-7
EINECS: 201-487-4
UN: 3077
ADR: 9,III
Order Now
Specification Test Results
Appearance Grey-brown to brown Crystalline powder
Infrared spectrum Conforms
HPLC ≥94 %