2,3-Dihydroxynaphthalene ≥98%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Naphthalene-2,3-diol, 2,3-Naphthalenediol
A11307.09 A11307.18 A11307.30
A11307.09EA 25.5 EUR
A11307.09 A11307.18 A11307.30
2,3-Dihydroxynaphthalene ≥98%
2,3-Dihydroxynaphthalene
Purity: 98% CAS No.: 92-44-4 Molecular Formula: C10H8O2 Formula Weight: 160.17 Flash Point: 175°(347°F) Melting Point: 162-165° Beilstein Reference: 742375 MDL No.: MFCD00004073
Formula: C₁₀H₈O₂
MW: 160.17 g/mol
Boiling Pt: 354
Melting Pt: 162…165 °C
Density: 1.12 g/cm³ (20 °C)
Flash Pt: 175 °C (347 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004073
CAS Number: 92-44-4
EINECS: 202-156-7