Naphthalene, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ200
147141-1KG 147141-5KG 147141-25G 147141-3KG
SIAL147141-1KGEA 53.8 EUR
SIAL147141-1KG SIAL147141-5KG SIAL147141-25G SIAL147141-3KG
Naphthalene, Sigma-Aldrich®
Naphthalene
Formula: C₁₀H₈
MW: 128.17 g/mol
Boiling Pt: 218
Melting Pt: 79…82 °C
Density: 1.145 g/cm³ (20 °C)
Flash Pt: 79
Storage Temperature: Ambient
MDL Number: MFCD00001742
CAS Number: 91-20-3
EINECS: 202-049-5
UN: 1334
ADR: 4.1 III
Merck Index: 12,06457
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c87c606cb828e0f39bab8b9d50c2c26501e1637d&vid=9e4bc21353014391fc8486208c8757e6375e2ef5&prid=web&clid=SAPDAM