5-Bromoacenaphthene

Supplier: Thermo Scientific

RH00779.1 RH00779.10 RH00779.50
MAYBRH00779.1EA 0 EUR
MAYBRH00779.1 MAYBRH00779.10 MAYBRH00779.50
5-Bromoacenaphthene
5-Bromoacenaphthene
Formula: C₁₂H₉Br
Boiling Pt: 335 °C (1013 hPa)
Melting Pt: 54 °C
Density: 1.1452 g/cm³ (20 °C)
MDL Number: MFCD00003809
CAS Number: 2051-98-1
EINECS: 218-138-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR