5-Iodoacenaphthene

Supplier: Thermo Scientific

RH00787.250 RH00787.1
MAYBRH00787.250EA 0 EUR
MAYBRH00787.250 MAYBRH00787.1
5-Iodoacenaphthene
5-Iodoacenaphthene
Formula: C₁₂H₉I
CAS Number: 6861-64-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR