Copper(I) bromide 97, TECHNICAL
Supplier: VWR Chemicals
Warning
23071.237EA 0 EUR
23071.237 23071.290
Copper(I) bromide 97, TECHNICAL
Copper (I) bromide
Formula: BrCu
MW: 143.45 g/mol
Boiling Pt: 1345
Melting Pt: 504
Density: 4.71 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010969
CAS Number: 7787-70-4
EINECS: 232-131-6
Merck Index: 13,02689
Order Now
Specification Test Results
Assay Min. 97 %