Copper(I) bromide, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ200
254185-100G 254185-10G
SIAL254185-100GEA 1220 EUR
SIAL254185-100G SIAL254185-10G
Copper(I) bromide, Sigma-Aldrich®
Copper (I) bromide
Formula: BrCu
Boiling Pt: 1345 °C (1013 hPa)
Melting Pt: 504 °C
Density: 4.71 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010969
CAS Number: 7787-70-4
EINECS: 232-131-6
Merck Index: 13,02689