Copper(II) chloride, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Copper dichloride, Cupric chloride
M-Clarity™ quality level = MQ100
451665-25G 451665-5G
SIAL451665-25GEA 279 EUR
SIAL451665-25G SIAL451665-5G
Copper(II) chloride, Sigma-Aldrich®
Copper (II) chloride
Formula: Cl₂Cu
MW: 134.45 g/mol
Boiling Pt: 1365.85
Melting Pt: 628
Density: 3.054 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010972
CAS Number: 7447-39-4
EINECS: 231-210-2
UN: 2802
ADR: 8 III
Merck Index: 13,02660