Silver chloride, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
204382-1G 204382-25G 204382-5G
SIAL204382-1GEA 30.9 EUR
SIAL204382-1G SIAL204382-25G SIAL204382-5G
Silver chloride, Sigma-Aldrich®
Silver chloride
Formula: AgCl
Boiling Pt: 1554 °C (1013 hPa)
Melting Pt: 455 °C
Density: 5.56 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003399
CAS Number: 7783-90-6
EINECS: 232-033-3
Merck Index: 13,08582