Silver cyanide, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
184535-10G 184535-50G
SIAL184535-10GEA 80.5 EUR
SIAL184535-10G SIAL184535-50G
Silver cyanide, Sigma-Aldrich®
Silver cyanide
Formula: AgCN
Melting Pt: 320…350 °C
Density: 3.943 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003409
CAS Number: 506-64-9
EINECS: 208-048-6
UN: 1684
ADR: 6.1,II
Merck Index: 12,08655