Silver fluoride, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ100

226866-25G 226866-5G
SIAL226866-25GEA 333 EUR
SIAL226866-25G SIAL226866-5G
Silver fluoride, Sigma-Aldrich®
Silver fluoride
Formula: AgF
Boiling Pt: 1150 °C (1013 hPa)
Melting Pt: 433 °C
Density: 5.85 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003410
CAS Number: 7775-41-9
EINECS: 231-895-8
UN: 3260
ADR: 8,II
Merck Index: 12,08657

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR