Copper(I) iodide 98%
Supplier: Thermo Fisher Scientific
Danger
194900010 194902500 194900050
ACRO194900010EA 266 EUR
ACRO194900010 ACRO194902500 ACRO194900050
Copper(I) iodide 98%
Copper (I) iodide
Formula: CuI
Boiling Pt: 1290 °C (1013 hPa)
Melting Pt: 605 °C
Density: 5,62 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010978
CAS Number: 7681-65-4
EINECS: 231-674-6
UN: 3077
ADR: 9,III
Merck Index: 13,02692
Order Now
Specification Test Results
Appearance Off-white to grey-brown Powder
Titration Complexometric ≥97.5 %
Sulfate (SO4) ≤5000 ppm
Iron (Fe) ≤50 ppm