Silver nitrate, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ200
204390-10G 204390-1G 204390-250G 204390-50G
SIAL204390-10GEA 91.2 EUR
SIAL204390-10G SIAL204390-1G SIAL204390-250G SIAL204390-50G
Silver nitrate, Sigma-Aldrich®
Silver nitrate
Formula: AgNO₃
Boiling Pt: 444 °C (1013 hPa)
Melting Pt: 210 °C
Density: 4.35 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003414
CAS Number: 7761-88-8
EINECS: 231-853-9
UN: 1493
ADR: 5.1,II
REACH: 01-2119513705-43
Merck Index: 12,08661
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM