Copper(II) oxide, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Cupric oxide, Copper oxide, Copper monoxide granulated
M-Clarity™ quality level = MQ100
450804-10G 450804-50G
SIAL450804-10GEA 247 EUR
SIAL450804-10G SIAL450804-50G
Copper(II) oxide, Sigma-Aldrich®
Copper (II) oxide
Formula: CuO
Boiling Pt: 2000 °C (1013 hPa)
Melting Pt: 1326 °C
Density: 4.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010979
CAS Number: 1317-38-0
EINECS: 215-269-1
UN: 3077
ADR: 9,III
Merck Index: 13,02674