Silver(I) oxide, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: di-Silver oxide, Disilver oxide, Silver oxide

M-Clarity™ quality level = MQ100

226831-1G 226831-25G 226831-5G
SIAL226831-1GEA 50.3 EUR
SIAL226831-1G SIAL226831-25G SIAL226831-5G
Silver(I) oxide, Sigma-Aldrich®
Silver(I) oxide
Formula: Ag₂O
MW: 231.74 g/mol
Melting Pt: 800…830 °C
Density: 7.14 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00003404
CAS Number: 20667-12-3
EINECS: 243-957-1
UN: 1479
ADR: 5.1 II
Merck Index: 12,08665

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR