Copper (I) sulphide, Sigma-Aldrich®
Supplier: Merck
Synonyms: Dicopper sulphide, Chacolcite, Copper(i)sulphide
M-Clarity™ quality level = MQ100
342459-1KG 342459-250G
SIAL342459-1KGEA 1050 EUR
SIAL342459-1KG SIAL342459-250G
Copper (I) sulphide, Sigma-Aldrich®
Copper (I) sulphide
Formula: Cu₂S
Melting Pt: 1100 °C
Density: 5.783 g/cm³ (25 °C)
MDL Number: MFCD00016062
CAS Number: 22205-45-4
EINECS: 244-842-9
Merck Index: 13,02697