Silver(I) sulphide
Supplier: Roth Carl
|
Warning
|
Synonyms:
Disilver sulphide
5311.1
5311.2
ROTH5311.1EA
0
EUR
ROTH5311.1
ROTH5311.2
Silver(I) sulphide
Silver (I) sulphide
Formula:
Ag₂S Melting Pt: 842 °C Density: 7.23 g/cm³ (25 °C) |
MDL Number:
MFCD00003406 CAS Number: 21548-73-2 EINECS: 244-438-2 Merck Index: 12,08677 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...